Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclooxygenase-1 Mouse anti-Human, Bovine, Mouse, Rat, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB5586
Description
Anti-Cyclooxygenase-1 Antibody is an antibody against Cyclooxygenase-1 for use in western blot, ELISA.
Specifications
Cyclooxygenase-1 | |
Monoclonal | |
Purified immunoglobulin. Liquid in 100mM Tris, pH 7.5 with 0.05% sodium azide. | |
PTGS1, PCOX1, PGHS1, COX-1, PGHS-1, PGG/HS, PHS1, COX1, PTGHS, COX3, EC 1.14.99.1 | |
Purified Sheep Cox-1. | |
Primary | |
−20°C in undiluted aliquots for 6 months. Avoid repeated freeze/thaw cycles. |
ELISA, Western Blot | |
Unconjugated | |
NM_000962.2; NM_080591.1 | |
Mouse | |
50 μg | |
Bovine, Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction