Learn More
CYP19A1, Mouse, Clone: 2B6, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CYP19A1.
Supplier: Abnova Corporation H00001588M03
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and catalyzes the last steps of estrogen biosynthesis, three successive hydroxylations of the A ring of androgens. Mutations in this gene can result in either increased or decreased aromatase activity; the associated phenotypes suggest that estrogen functions both as a sex steroid hormone and in growth or differentiation. The gene expresses two transcript variants. [provided by RefSeq
Sequence: PGPGYCMGIGPLISHGRFLWMGIGSACNYYNRVYGEFMRVWISGEETLIISKSSSMFHIMKHNHYSSRFGSKLGLQCIGMHEKGIIFNNNPELWKTTRPFFMKALSGPGLVRMVTVCAESLKTHLDRLEEVTNESGYVDVLTLLRRVMLDTSNTLFLRIPLDGTEIFTLTSSpecifications
| CYP19A1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC035714 | |
| CYP19A1 | |
| CYP19A1 (AAH35714, 48 a.a. ∼ 218 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 2B6 | |
| Mouse monoclonal antibody raised against a full length recombinant CYP19A1. | |
| CYP19A1 | |
| ARO/ARO1/CPV1/CYAR/CYP19/MGC104309/P-450AROM | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1588 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.