Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CYP19A1, Polyclonal

Catalog No. ST1564100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1564100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1564100UG Supplier MilliporeSigma™ Supplier No. ST1564100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects CYP19A1 Clone: in Human samples, and it is validated for Immuno Blotting

CYP19A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and catalyzes the last steps of estrogen biosynthesis, three successive hydroxylations of the A ring of androgens. Mutations in this gene can result in either increased or decreased aromatase activity; the associated phenotypes suggest that estrogen functions both as a sex steroid hormone and in growth or differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP19A1
Applications Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Rabbit
Immunogen Full-length, human CYP19A1 (aa 1-503)
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the CYP19A1 protein in human liver tissue.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.