Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP24A1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABN201MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN201MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN201MI Supplier MilliporeSigma Supplier No. ABN201
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-CYP24A1 Antibody is an antibody against CYP24A1 for use in 0.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP24A1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols CYP24A1; CYP24
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human CYP24A1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_000773
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.