Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cytochrome P450, family 24, subfamily A, polypeptide 1, Mouse, Clone: 1F8, Abnova™

Catalog No. 89014087 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-087 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-087 Supplier Abnova Corporation Supplier No. H00001591M07
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CYP24A1.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This mitochondrial protein initiates the degradation of 1,25-dihydroxyvitamin D3, the physiologically active form of vitamin D3, by hydroxylation of the side chain. In regulating the level of vitamin D3, this enzyme plays a role in calcium homeostasis and the vitamin D endocrine system. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: LMLNTQVLGSSEDNFEDSSQFRPERWLQEKEKINPFAHLPFGVGKRMCIGRRLAELQLHLALCWIVRKYDIQATDNEPVEMLHSGTLVPSRELPIAFCQR

Specifications

Antigen cytochrome P450, family 24, subfamily A, polypeptide 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CYP24A1.
Formulation PBS with no preservative; pH 7.4
Gene CYP24A1
Gene Accession No. NM_000782
Gene Alias CP24/CYP24/MGC126273/MGC126274/P450-CC24
Gene Symbols CYP24A1
Host Species Mouse
Immunogen CYP24A1 (NP_000773, 415 a.a. ∼ 514 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1591
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.