Learn More
CYP2A13, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human CYP2A13 peptide using ARM Technology.
Supplier: Abnova Corporation H00001553K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum. Although its endogenous substrate has not been determined, it is known to metabolize 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone, a major nitrosamine specific to tobacco. This gene is part of a large cluster of cytochrome P450 genes from the CYP2A, CYP2B and CYP2F subfamilies on chromosome 19q. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
CYP2A13 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
CYP2A13 | |
CYP2A13 | |
A synthetic peptide of human CYP2A13 | |
RUO | |
1553 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
cytochrome P450, family 2, subfamily A, polypeptide 13 | |
CYP2A13 | |
CPAD, CYP2A | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.