Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP2b10 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB9916MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9916MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9916MI Supplier MilliporeSigma Supplier No. AB9916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-CYP2b10 Antibody is validated for use in western blotting for the detection of CYP2b10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2b10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Raw rabbit serum with 0.05% sodium azide.
Gene Symbols EC 1.14.14.1; CYPIIB10; P450-16-alpha; Cytochrome P450 2B20; CYPIIB20
Host Species Rabbit
Immunogen Recombinant 2b10
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_009998
Target Species Mouse
Content And Storage −20°C for 12 months
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.