Learn More
CYP2E1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CYP2E1.
Supplier: Abnova Corporation H00001571A01
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is induced by ethanol, the diabetic state, and starvation. The enzyme metabolizes both endogenous substrates, such as ethanol, acetone, and acetal, as well as exogenous substrates including benzene, carbon tetrachloride, ethylene glycol, and nitrosamines which are premutagens found in cigarette smoke. Due to its many substrates, this enzyme may be involved in such varied processes as gluconeogenesis, hepatic cirrhosis, diabetes, and cancer. [provided by RefSeq
Sequence: DSVLYDNQEFPDPEKFKPEHFLNENGKFKYSDYFKPFSTGKRVCAGEGLARMELFLLLCAILQHFNLKPLVDPKDIDLSPIHIGFGCIPPRYKLCVIPRSSpecifications
CYP2E1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant CYP2E1. | |
CYP2E1 | |
CPE1/CYP2E/P450-J/P450C2E | |
Mouse | |
50 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_000773 | |
CYP2E1 | |
CYP2E1 (NP_000764, 394 a.a. ∼ 493 a.a) partial recombinant protein with GST tag. | |
RUO | |
1571 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.