Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP450 1A1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB10088MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10088MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10088MI Supplier MilliporeSigma Supplier No. AB10088
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-CYP450 1A1 Antibody is validated for use in western blotting for the detection of CYP450 1A1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP450 1A1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Raw Rabbit Serum with 0.1% Sodium Azide
Gene Symbols CYP1A1; CP11; P1-450; CYPIA1; P450-C; CYP1; P450DX; P450-P1; AHRR; AHH
Host Species Rabbit
Immunogen Recombinant CYP450 1A1
Purification Method Serum
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000499.2
Target Species Human
Content And Storage 2-8°C for 12 months
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.