Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP46A1, Mouse, Clone: 2B5, Abnova™

Catalog No. 89002777 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-777 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-777 Supplier Abnova Corporation Supplier No. H00010858M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CYP46A1.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein is expressed in the brain, where it converts cholesterol to 24S-hydroxycholesterol. While cholesterol cannot pass the blood-brain barrier, 24S-hydroxycholesterol can be secreted in the brain into the circulation to be returned to the liver for catabolism. [provided by RefSeq

Sequence: TSMLLGAQKPLSQAVKLMLEGITASRNTLAKFLPGKRKQLREVRESIRFLRQVGRDWVQRRREALKRGEEVPADILTQILKAEEGAQDDEGLLDNFVTFF

Specifications

Antigen CYP46A1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2B5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CYP46A1.
Formulation PBS with no preservative; pH 7.4
Gene CYP46A1
Gene Accession No. NM_006668
Gene Alias CP46/CYP46
Gene Symbols CYP46A1
Host Species Mouse
Immunogen CYP46A1 (NP_006659, 201 a.a. ∼ 300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10858
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.