Learn More
CYP4Z1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human CYP4Z1 peptide using ARM Technology.
Supplier: Abnova Corporation H00199974K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This gene is part of a cluster of cytochrome P450 genes on chromosome 1p33. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
CYP4Z1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
CYP4Z1 | |
CYP4Z1 | |
A synthetic peptide of human CYP4Z1 | |
RUO | |
199974 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
cytochrome P450, family 4, subfamily Z, polypeptide 1 | |
CYP4Z1 | |
CYP4A20 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.