Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyp7a1 Mouse anti-Human, Rat, Mouse, Clone: 15B9.1, MilliporeSigma™

Catalog No. MABD42MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD42MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD42MI Supplier MilliporeSigma Supplier No. MABD42
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Cyp7a1 antibody is validated for use in western blotting and immunohistochemistry for the detection of the Cyp7a1 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyp7a1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 15B9.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000771
Gene Symbols CYP7A1, CYP7
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Cyp7a1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.