Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cysteine-rich Motor Neuron 1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB5699MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5699MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5699MI Supplier MilliporeSigma Supplier No. AB5699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Cysteine-rich Motor Neuron 1 Antibody detects level of Cysteine-rich Motor Neuron 1 and has been published and validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cysteine-rich Motor Neuron 1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Liquid.
Gene Symbols CRIM1; CRIM-1; MGC138194; S52
Host Species Rabbit
Immunogen Recombinant protein from the cytoplasmic tail of mouse Crim1.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_016441.1
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.