Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cysteine String Protein Rabbit anti-All, Polyclonal, MilliporeSigma™

Catalog No. AB1576MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1576MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1576MI Supplier MilliporeSigma Supplier No. AB1576

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

This Anti-Cysteine String Protein Antibody is validated for use in western blot, immunocytochemistry, immunohistochemistry for the detection of Cysteine String Protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cysteine String Protein
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid rabbit serum containing no preservatives
Gene Symbols DNAJC5; CSP; DKFZP434N1429; FLJ13070; DKFZp761N1221; FLJ00118
Host Species Rabbit
Immunogen Recombinant rat cysteine-string protein with the cysteine string deleted.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_025219.1
Target Species All species
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.