Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cystic Fibrosis Transmembrane Conductance Regulator Mouse anti-Human, Clone: L12B4, MilliporeSigma™

Catalog No. MAB3484MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3484MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3484MI Supplier MilliporeSigma Supplier No. MAB3484
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Cystic Fibrosis Transmembrane Conductance Regulator Antibody, a.a. 386-412, clone L12B4 is an antibody against Cystic Fibrosis Transmembrane Conductance Regulator for use in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cystic Fibrosis Transmembrane Conductance Regulator
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone L12B4
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02MPhosphate buffer, 0.25MNaCl, pH 7.6 with 0.1% sodium azide.
Gene Accession No. NM_000492.3
Gene Symbols CFTR, ABCC7, MRP7, TNR-CFTR, CFTR/MRP, CBAVD, dJ760C5.1, CF, ABC35
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.