Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome c Mouse, Unlabeled, Clone: 7H8.2C12, BD

Catalog No. BDB556433 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556433 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556433 Supplier BD Biosciences Supplier No. 556433
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A cytochrome is an electron-transporting protein that contains a heme prosthetic group. Cytochromes have been known to be essential components of the mitochondrial respiratory chain since 1925. The iron atom of the heme group in cytochromes alternates between a reduced ferrous (+2) state and an oxidized ferric (+3) state during electron transport in oxidative phosphorylation. Cytochromes are classified into four groups (a, b, c, and d) according to spectrochemical characteristics, and there are five cytochromes between Coenzyme QH2 and O2 in the electron transport chain. Cytochrome c is a water-soluble peripheral membrane protein of the mitochondria and transports electrons from the Coenzyme QH2 - Cytochrome c reductase complex to the Cytochrome c oxidase complex. Recently it has been suggested that Cytochrome c may be required for cells to undergo apoptosis. Mitochondria may be involved in apoptosis by releasing Cytochrome c to the cytosol where it functions in the activation of Caspase 3 (CPP32), an ICE family molecule that has been shown to trigger apoptosis when activated. In the cytoplasm Cytochrome C might exist in polymeric forms, appearing as a 58-60kDa band by western blot analysis, rather than the monomeric form. Monomeric Cytochrome c migrates at a reduced molecular weight of 15kDa. Clone 7H8.2C12 recognizes the denatured form of pigeon, horse, rat, mouse and human Cytochrome c. It does not recognize the native form of Cytochrome c. Clone 7H8.2C12 recognizes an epitope within amino acids 93-104 (inclusive) of pigeon Cytochrome C, based on competitive ELISA results. Synthetic peptides corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon Cytochrome c were used as immunogens. Clone 7H8.2C12 is used for western blot detection of Cytochrome c. Clone 6H2.B4, Mfr. No. 556432, is used for immunoprecipitation. Cell lines P388D1 mouse lymphoma (ATCC CCL-46), HeLa human carcinoma (ATCC CCL-2), Jurkat T leukemia (ATCC TIB-152), and NIH/3T3 mouse fibroblast cells (ATCC CRL-1658) may be used as positive control cell lysates. This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 7H8.2C12
Isotype: IgG2b, κ
Species Reactivity: Human
Immunogen: Synthetic Peptides of Pigeon Cytochrome C

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome c
Applications Western Blot
Classification Monoclonal
Clone 7H8.2C12
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Synthetic Peptides of Pigeon Cytochrome C
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Pigeon, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.