Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome C Mouse anti-Human, Mouse, Rat, Clone: 7H8.2C12, MilliporeSigma™

Catalog No. MAB1800MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1800MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1800MI Supplier MilliporeSigma Supplier No. MAB1800

Mouse Monoclonal Antibody

This Anti-Cytochrome C Antibody, clone 7H8.2C12 is validated for use in western blotting for the detection of Cytochrome C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome C
Applications Western Blot
Classification Monoclonal
Clone 7H8.2C12
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS containing 0.5% BSA, 0.01% Thimerosal and 50% glycerol.
Gene Accession No. NM_018947.4
Gene Symbols CYCS, CYC, HCS
Host Species Mouse
Immunogen Pigeon Cytochrome C
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.