Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome C Sheep anti-Canine, Human, Rat, Rabbit, Polyclonal, MilliporeSigma™

Catalog No. AB3547MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3547MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3547MI Supplier MilliporeSigma Supplier No. AB3547
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Anti-Cytochrome C Antibody is an antibody against Cytochrome C for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome C
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified sheep immunoglobulin. Liquid in PBS containing 0.08% sodium azide.
Gene Symbols CYCS; CYC; HCS
Host Species Sheep
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_018947.4
Target Species Canine, Human, Rabbit, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.