Learn More
Invitrogen™ Cytochrome C Polyclonal Antibody

Chicken Polyclonal Antibody
Supplier: Invitrogen™ PA19585
Description
PA1-9585 detects Cytochrome C in human, mouse, bovine and rat samples. PA1-9585 has been successfully used in Western blot and ELISA procedures. By Western blot, PA1-9585 detects a 12 kDa protein representing Cytochrome C. PA1-9585 immunogen is a purified bovine cytochrome c protein.
Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.
Specifications
Cytochrome C | |
Polyclonal | |
Unconjugated | |
Cycs | |
CG17903; CG17903-PA; CG17903-PB; CRF; CYC; CYC2; Cycs; CYCSA; Cyct-c-p; Cyt c; cyt c-p; cyt.c; CytC; Cyt-c; CYTC2; Cyt-c2; CytC-2; cyt-c-p; cytC-p; Cyt-c-p-PA; Cyt-c-p-PB; Cytochrome c; cytochrome c DC4; Cytochrome c proximal; Cytochrome C, expressed in somatic tissues; cytochrome c, somatic; Cytochrome c-2; cytochrome c-like; cytochrome c-like protein; Cytochrome c-proximal; cytochrome-c-p; Cytochrome-c-proximal; DC4; dCyt.C-p; DMc01; Dmel\CG17903; Dmel_CG17903; hc1; HCS; l(2)k13905; lethal (2) k13905; LOC100341175; LOC100357840; MGC128356 protein; pDMc01; RH17228p; THC4 | |
Chicken | |
Affinity Chromatography | |
RUO | |
13063, 25309, 510767, 54205 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P62894, P62897, P62898, P99999 | |
Cycs | |
Purified bovine cytochrome c protein. | |
100 μg | |
Primary | |
Human, Mouse, Rat, Bovine | |
Antibody | |
IgY |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.