Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Cytochrome P450 3A4, Clone: EPR6202, Rabbit Monoclonal,

Catalog No. MABS756MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS756MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS756MI Supplier MilliporeSigma™ Supplier No. MABS756

Specifications

Antigen Cytochrome P450 3A4
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone EPR6202
Conjugate Unconjugated
Formulation Rabbit Monoclonal in buffer containing PBS with up to 0.1% sodium azide, 0.05% BSA, and 50% glycerol.
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminus of human Cytochrome P450 3A4.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) CYP3A4, CYP3A3
Target Species Human
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.