Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 3A4 Mouse anti-Human, Clone: F24 P2 B10, MilliporeSigma™

Catalog No. MAB10054MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10054MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB10054MI Supplier MilliporeSigma Supplier No. MAB10054

Mouse Monoclonal Antibody

This Anti-Cytochrome P450 3A4 Antibody, clone F24 P2 B10 is validated for use in western blotting for the detection of Cytochrome P450 3A4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 3A4
Applications Western Blot
Classification Monoclonal
Clone F24 P2 B10
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_059488
Gene Symbols CP33, CP34, CYP3A, CYP3A3, CYPIIIA3, CYPIIIA4, HLP, HLp, NF-25, P450-PCN1, P450C3, P450PCN1
Host Species Mouse
Immunogen Ovalbumin-conjugated linear peptide corresponding to human Cytochrome P450 3A4.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.