Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 Aldosterone Synthase Mouse anti-Mouse, Rat, Clone: 1G5, MilliporeSigma™

Catalog No. MAB6021MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6021MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB6021MI Supplier MilliporeSigma Supplier No. MAB6021
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Cytochrome P450 Aldosterone Synthase Antibody, clone 1G5 is validated for use in ELISA, immunocytochemistry, immunohistochemistry (paraffin), western blotting for the detection of Cytochrome P450 Aldosterone Synthase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 Aldosterone Synthase
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1G5
Conjugate Unconjugated
Formulation Unpurified tissue culture supernatant containing 50% glycerol and 0.1% sodium azide as a preservative.
Host Species Mouse
Immunogen Aldo-1-MAP (aa 175-192) (KVRQNARGSLTMDVQQSL-MAP)
Quantity 500 μL
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.