Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 CYP 2C6 Mouse anti-Human, Mouse, Rat, MilliporeSigma™

Catalog No. MAB10040MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10040MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB10040MI Supplier MilliporeSigma Supplier No. MAB10040

Mouse Monoclonal Antibody

Detect Cytochrome P450 CYP 2C6 using this Mouse Anti-Rat Cytochrome P450 CYP 2C6 Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 CYP 2C6
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Liquid containing 0.1% sodium azide.
Gene Accession No. XM_001081924.1
Host Species Mouse
Immunogen Rat liver cytochrome P450 2C6
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.