Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 Enzyme CYP2E1 Rabbit anti-Human, Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB1252MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1252MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1252MI Supplier MilliporeSigma Supplier No. AB1252
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Cytochrome P450 Enzyme CYP2E1 Antibody is an antibody against Cytochrome P450 Enzyme CYP2E1 for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 Enzyme CYP2E1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum in buffer containing no preservative.
Gene Symbols CYP2E1; OTTHUMP00000046571; CYPIIE1; CYP2E; P450C2E; P450-J; CPE1
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_000764
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.