Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 Enzyme CYP3A1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1253
Description
This Anti-Cytochrome P450 Enzyme CYP3A1 Antibody is validated for use in immunohistochemistry, western blotting for the detection of Cytochrome P450 Enzyme CYP3A1.
Specifications
Cytochrome P450 Enzyme CYP3A1 | |
Polyclonal | |
Rabbit antiserum. Liquid. Contains no preservative. | |
Synthetic peptide | |
Primary | |
Rat | |
Serum |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
100 μL | |
NP_037237.2 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction