Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 Enzyme CYP3A4 Rabbit anti-Human, Pig, Polyclonal, MilliporeSigma™

Catalog No. AB1254MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1254MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1254MI Supplier MilliporeSigma Supplier No. AB1254
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Cytochrome P450 Enzyme CYP3A4 Antibody detects level of Cytochrome P450 Enzyme CYP3A4 and has been published and validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 Enzyme CYP3A4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit antiSerum. Liquid containing no preservative.
Gene Symbols CYP3A4; CYPIIIA3; P450-PCN1; P450C3; P450PCN1; MGC126680; HLp; CYP3A; CP34; CYPIIIA4; CP33; NF-25; HLP; CYP3A3; EC 1.14.13.67; EC 1.14.13.97
Host Species Rabbit
Immunogen A synthetic peptide containing the C-terminal sequence of TVSGA from human CYP3A4. The peptide was coupled to KLH.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_017460.3
Target Species Human, Pig
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.