Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytohesin-1 Rat anti-Human, Clone: 7H2, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABT14
Description
This Anti-Cytohesin-1 Antibody, clone 7H2 is validated for use in western blot, immunofluoresence, immunoprecipitation for the detection of Cytohesin-1.
Specifications
Cytokeratin 1 | |
Monoclonal | |
Unconjugated | |
NP_004753 | |
Rat | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunofluorescence, Immunoprecipitation, Western Blot | |
7H2 | |
Purified rat monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
B2-1, PSCD1, CYTH1, SEC7 | |
Histidine-tagged recombinant protein corresponding to human Cytohesin-1. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction