Learn More
Cytokeratin 14, 15, 16 and 19 Mouse anti-Human, Alexa Fluor 647, Clone: KA4, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 563648
Description
The KA4 monoclonal antibody specifically binds to cytokeratins 14, 15, 16 and 19 present in the cytoplasm of a wide variety of human simple epithelium. Human epithelium has been shown to contain cytoplasmic filaments of the cytokeratin type. There have been 19 different polypeptides identified and they demonstrate a characteristic pattern for each type of epithelium. This antibody is useful for intracellular staining and flow cytometric analysis, Western blot and immunohistochemical staining of both, acetone-fixed frozen tissue sections and formalin-fixed paraffin embedded tissue sections with citrate pretreatment.

Specifications
Cytokeratin 14, 15, 16 and 19 | |
Monoclonal | |
Alexa Fluor 647 | |
Mouse | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
KA4 | |
KRT14, KRT15, KRT16, KRT19; CK14, CK15, CK16, CK19; K14, K15, K16, K19 | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.