Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 14, 15, 16 and 19 Set Mouse anti-Human, PE, Clone: KA4, BD

Catalog No. BDB550953 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550953 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550953 Supplier BD Biosciences Supplier No. 550953
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with cytokeratins 14, 15, 16 and 19 present in the cytoplasm of a wide variety of human simple epithelium. Human epithelium has been shown to contain cytoplasmic filaments of the cytokeratin type. There have been 19 different polypeptides identified and they demonstrate a characteristic pattern for each type of epithelium. This reagent is useful for intracellular staining for flow cytometric analysis, western blot and immunohistochemical staining of both, acetone-fixed frozen tissue sections and formalin-fixed paraffin embedded tissue sections with citrate pretreatment.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 14, 15, 16 and 19 Set
Applications Flow Cytometry
Classification Monoclonal
Clone KA4
Conjugate PE
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Includes PE Mouse Anti-Human Cytokeratin (100 tests), PE Mouse IgG1 Isotype Control (100 tests)
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.