Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 14 Mouse anti-Human, Clone: LL002, MilliporeSigma™

Catalog No. CBL197MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL197MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL197MI Supplier MilliporeSigma Supplier No. CBL197
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Cytokeratin 14 Antibody, clone LL002 is an antibody against Cytokeratin 14 for use in western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 14
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone LL002
Conjugate Unconjugated
Formulation The monoclonal is presented at a concentration of 100μg/mL in phosphate buffered saline containing 10mM sodium azide and 1% bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular applicati
Gene Accession No. NM_000526.3
Gene Symbols KRT14, Cytokeratin-14, NFJ, CK-14, CK14, EBS3, EBS4, K14, Keratin-14
Host Species Mouse
Immunogen Thyroglobulin conjugated synthetic peptide of extreme carboxy terminus of human keratin 14 (last 15 residues)
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C for up to 1 month, −20°C in undiluted aliquots for long term
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.