Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 14 Mouse anti-Canine, Human, Pig, Rat, Clone: RCK107, MilliporeSigma™

Catalog No. MAB3232MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3232MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3232MI Supplier MilliporeSigma Supplier No. MAB3232
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Cytokeratin 14 using this Anti-Cytokeratin 14 Antibody, clone RCK107 validated for use in flow cytometry, western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 14
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone RCK107
Conjugate Unconjugated
Formulation Liquid in buffer with 0.1% sodium azide.
Gene Accession No. NM_000526.3
Gene Symbols KRT14, Cytokeratin-14, NFJ, CK-14, CK14, EBS3, EBS4, K14, Keratin-14
Host Species Mouse
Immunogen Cytoskeletal preparation of TR146 epithelial cells.
Quantity 100 μg
Primary or Secondary Primary
Target Species Canine, Human, Pig, Rat
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.