Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 18 Mouse anti-Human, Clone: CK2, MilliporeSigma™

Catalog No. MAB3404CMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3404CMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3404CMI Supplier MilliporeSigma Supplier No. MAB3404C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Cytokeratin 18 Antibody, clone CK2 (Ascites Free) is validated for use in immunohistochemistry, western blotting and immunofluorescence for the detection of Cytokeratin 18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 18
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone CK2
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000215
Gene Symbols KRT18, CYK18, PIG46
Host Species Mouse
Immunogen Cytoskeletal preparation of HeLa cells expressing human Cytokeratin 18.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.