Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 18 Mouse anti-Human, Clone: CK2, MilliporeSigma™

Catalog No. MAB3404MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3404MI 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3404MI Supplier MilliporeSigma Supplier No. MAB3404
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Cytokeratin 18 Antibody, clone CK2 is validated for use in immunocytochemistry, immunohistochemistry for the detection of Cytokeratin 18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 18
Applications Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone CK2
Conjugate Unconjugated
Formulation Liquid in 0.02MPhosphate buffer, 0.25MNaCl with 0.1% sodium azide.
Gene Accession No. NM_000224.2; NM_199187.1
Gene Symbols KRT18, K18, Keratin-18, CYK18, Cytokeratin-18, CK-18
Host Species Mouse
Immunogen Cytoskeletal preparation of HeLa cells (Debus et al., 1982).
Quantity 40 μg
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, lyophilized antibody. 2-8°C, reconstitured antibody for up to 12 months.Do not Freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.