Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 18 Mouse anti-Human Only, Clone: RCK106, MilliporeSigma™

Catalog No. MAB3236MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3236MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3236MI Supplier MilliporeSigma Supplier No. MAB3236
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Cytokeratin 18 Antibody, clone RCK106 detects level of Cytokeratin 18 and has been published and validated for use in flow cytometry, western blot, immunocytochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 18
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone RCK106
Conjugate Unconjugated
Formulation Liquid in buffer with 0.1% sodium azide.
Gene Accession No. NM_000224.2; NM_199187.1
Gene Symbols KRT18, K18, Keratin-18, CYK18, Cytokeratin-18, CK-18
Host Species Mouse
Immunogen Cytoskeletal preparation of human bladder carcinoma cell line T24.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.