Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 19 Mouse anti-Human, Clone: RCK108, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB3238
Description
Anti-Cytokeratin 19 Antibody, clone RCK108 is an antibody against Cytokeratin 19 for use in flow cytometry, western blot, immunocytochemistry, immunohistochemistry (paraffin).
Specifications
Cytokeratin 19 | |
Monoclonal | |
Unconjugated | |
NM_002276.4 | |
Mouse | |
100 μg | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
RCK108 | |
Liquid in buffer with 0.1% sodium azide. | |
KRT19, Keratin-19, K19, CK19, MGC15366, K1CS, Cytokeratin-19, CK-19 | |
Cytoskeletal preparation of T24 cells. | |
Primary | |
−20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction