Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 20 Mouse anti-Human, Clone: IT-Ks20.8, MilliporeSigma™

Catalog No. CBL208MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL208MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL208MI Supplier MilliporeSigma Supplier No. CBL208
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Cytokeratin 20 using this Anti-Cytokeratin 20 Antibody, clone IT-Ks20.8 validated for use in western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 20
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone IT-Ks20.8
Conjugate Unconjugated
Formulation Protein A purified immunoglobulin lyophilized from PBS, pH 7.4, containing 0.5% BSA and 0.09% sodium azide. Reconstitute with 1 mL distilled water.
Gene Accession No. NM_019010.1
Gene Symbols KRT20, K20, CK20, Keratin-20, CK-20, KRT21, Cytokeratin-20, CD20, MGC35423
Host Species Mouse
Immunogen Electrophoretically purified cytokeratin 20 from human intestinal mucosa
Quantity 50 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.