Learn More
MilliporeSigma™ anti-Cytokeratin 5, 6, Clone: D5/16B4, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB1620/DEL

Description
The cytokeratins are a family of intermediate filaments fond in almost all epithelial cells. They are divided into four subclasses depending on their patterns of reactivity and electrophoretic characteristics and are also arranged I a numbered series of 20 or more acoring to molecular weight and isoelectric point (Quinlan et al. 1985). Types 5 and 6 of cytokeratin (CK5/6) are in high frequency in mesothelial cells, as opposed to cells of adenocarcinoma. CK5/6 has been shown to be a reliable marker for mesothelioma and squamous cell carcinoma of the lung and Bowen's disease. It does not react with pulmonary adenocarcinoma.
Wild & Mischke, 1986; Mischke & Wild, 1987
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting

Specifications
Cytokeratin 5, 6 | |
Unconjugated | |
Purified cytokeratin 5 | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
50 μg | |
Cell Structure | |
Human, Mouse, Rat | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.