Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 5 Mouse anti-Human, Mouse, Rat, Clone: D5/16B4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1620
Description
Anti-Cytokeratin 5 Antibody, 6, clone D5/16B4 is an antibody against Cytokeratin 5 for use in immunofluoresence, immunohistochemistry (paraffin) and western blotting.
Specifications
Cytokeratin 5 | |
Monoclonal | |
Unconjugated | |
NM_000424.3 | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 6 months. | |
IgG1 |
Immunofluorescence, Immunohistochemistry, Western Blot | |
D5/16B4 | |
Purified mouse monoclonal IgG1 in buffer containing 0.02MPhosphate buffer, 0.25MNaCl, pH 7.6 with 0.1% sodium azide. | |
KRT5, K5, EBS2, DDD, Keratin-5, KRT5A, Cytokeratin-5, CK5, CK-5 | |
Purified cytokeratin 5 (Exp. Cell Res. (1986) 162:114; J. Invest. Dermatology (1987) 88:191). | |
50 μg | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction