Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 7 Mouse anti-Human, Clone: OV-TL 12/30, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB3554
Description
Anti-Cytokeratin 7 Antibody, clone OV-TL 12/30 detects level of Cytokeratin 7 and has been published and validated for use in flow cytometry, western blot, immunocytochemistry, immunohistochemistry (paraffin).
Specifications
Cytokeratin 7 | |
Monoclonal | |
Unconjugated | |
NM_019010.1 | |
Mouse | |
100 μg | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
OV-TL 12/30 | |
Liquid in PBS containing 0.09% sodium azide. | |
KRT20, Cytokeratin-20, K20, CK20, CD20, MGC35423, Keratin-20, CK-20, KRT21 | |
Cytoskeletal preparation of OTN-11 ovarian carcinoma cell line. | |
Primary | |
−20°C in undiluted aliquots for 6 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction