Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 7 Mouse, Unlabeled, Clone: RCK105, BD

Catalog No. BDB550507 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550507 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550507 Supplier BD Biosciences Supplier No. 550507

Mouse Monoclonal Antibody

Intermediate filaments (IF) are a subset of cytoskeletal proteins which function to give overall structural integrity to the plasma membrane as well as organize cells into specific tissues. IF proteins can be divided into six major types based upon the similarity in sequence. Type I and II IF proteins are designated keratins based upon whether they are acidic (I) or basic (II) and are characterized by their remarkable biochemical diversity. A subgroup of keratins, designated cytokeratins, are represented in epithelial tissues by at least 20 different polypeptides, designated
1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin 5 has a molecular weight of approximately 58kDa (NCBI Accession number P13647) while cytokeratin 8 is approximately 53kDa (NCBI Accession number P05787). We observe cytokeratin 5 to migrate at approximately 54kDa and cytokeratin 8 at approximately 52kDa. The antibody recognizes cytokeratins 5 and 8 from human, mouse, rat, hamster, rabbit, canine, and swine. The antibody stains all epithelial tissue; it does not stain non-epithelial tissue. The human lung cancer cell line (MR21) was used as the immunogen.

Host Species: Mouse
Clone: RCK102
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human lung cancer cell line (MR21)

Immunohistochemistry (Frozen), Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 7
Applications Western Blot
Classification Monoclonal
Clone RCK105
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human lung cancer cell line (MR21)
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Hamster, Human, Mouse, Pig, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.