Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 7 Mouse anti-Canine, Human, Hamster, Mouse, Pig, Rat, Feline, Clone: RCK105, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB3226
Description
Anti-Cytokeratin 7 Antibody, clone RCK105 is an antibody against Cytokeratin 7 for use in flow cytometry, western blot, immunocytochemistry, immunohistochemistry.
Specifications
Cytokeratin 7 | |
Monoclonal | |
Unconjugated | |
NM_019010.1 | |
Mouse | |
100 μg | |
Canine, Feline, Hamster, Human, Mouse, Pig, Rat | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
RCK105 | |
Liquid in buffer with 0.1% sodium azide. | |
KRT20, K20, CK20, Keratin-20, CK-20, KRT21, Cytokeratin-20, CD20, MGC35423 | |
Cytokeratins from the human bladder carcinoma cell line T24. | |
Primary | |
−20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction