Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin AE1/AE3 Mouse anti-Human, Rat, Mouse, Monkey, Rabbit, Bovine, Chicken, Clone: AE1/AE3, MilliporeSigma™

Catalog No. MAB3412MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3412MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3412MI Supplier MilliporeSigma Supplier No. MAB3412
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Cytokeratin AE1/AE3 Antibody, recognizes acidic and basic cytokeratins, clone AE1/AE3 is an antibody against Cytokeratin AE1/AE3 for use in ELISA, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin AE1/AE3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone AE1/AE3
Conjugate Unconjugated
Formulation Anti-keratin AE1/AE3 is supplied in 0.5 mL borate-buffered saline, pH 8.0, containing 0.09% sodium azide
Host Species Mouse
Immunogen Human epidermal keratins
Purification Method Sodium sulfate precipitation
Quantity 500 μg
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Monkey, Mouse, Rabbit, Rat
Content And Storage 2-8°C for 12 months. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.