Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IHC Select™ Cytokeratin AE1/AE3 (Pan cytokeratins) Mouse anti-Human, Clone: AE1/AE3, MilliporeSigma™

Catalog No. IHCR20256MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
6 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IHCR20256MI 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IHCR20256MI Supplier MilliporeSigma Supplier No. IHCR20256
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

immunohistochemistry Select Anti-Cytokeratin AE1/AE3 (Pan cytokeratins) Antibody, prediluted, clone AE1/AE3 is an antibody against Cytokeratin AE1/AE3 (Pan cytokeratins) for use in immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin AE1/AE3 (Pan cytokeratins)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AE1/AE3
Conjugate Unconjugated
Formulation Purified monoclonal mouse antibody supplied in liquid format diluted in PBS, pH 7.2 with stabilizers, 0.2% Tween 20, and 0.1% ProClin 300 as preservative.
Host Species Mouse
Immunogen Human epidermal keratin
Quantity 6 mL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.