Learn More
Cytokeratin Mouse anti-Human, Brilliant Violet 510, Clone: CAM 5.2, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 563613
Description
The CAM5.2 monoclonal antibody specifically recognizes cytokeratin having a primary reactivity with human keratin proteins that correspond to Moll's peptides #7 (48 kDa) and #8 (52 kDa). Cytokeratin 7 and 8 are type II cytoskeletal keratins. These cytoskeletal proteins provide structural integrity for epithelial cells and may serve other functions as well. They are expressed in epithelia cells that comprise normal human tissues. Although these cytokeratins are not normally expressed in stratified squamous epithelium, they may be expressed in some squamous cell carcinomas. The CAM 5.2 antibody stains most epithelial-derived tissue, including liver, renal tubular epithelium, and hepatocellular and renal cell carcinomas.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon™ BV510 can be excited by the violet laser and detected in the BD Horizon™ V500 (525/50-nm) filter set. BD Horizon™ BV510 conjugates are useful for the detection of dim markers off the violet laser.
Specifications
| Cytokeratin | |
| Monoclonal | |
| Brilliant Violet 510 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human HT29 colorectal carcinoma cell line | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| CAM 5.2 | |
| Cytokeratin-7/-8; CK-7/CK-8; | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.