Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin Mouse anti-Human, Alexa Fluor 647, Clone: CAM 5.2, BD

Catalog No. BDB563614 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563614 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563614 Supplier BD Biosciences Supplier No. 563614
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CAM5.2 monoclonal antibody specifically recognizes cytokeratin having a primary reactivity with human keratin proteins that correspond to Moll's peptides #7 (48 kDa) and #8 (52 kDa). Cytokeratin 7 and 8 are type II cytoskeletal keratins. These cytoskeletal proteins provide structural integrity for epithelial cells and may serve other functions as well. They are expressed in epithelia cells that comprise normal human tissues. Although these cytokeratins are not normally expressed in stratified squamous epithelium, they may be expressed in some squamous cell carcinomas. The CAM 5.2 antibody stains most epithelial-derived tissue, including liver, renal tubular epithelium, and hepatocellular and renal cell carcinomas.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin
Applications Flow Cytometry
Classification Monoclonal
Clone CAM 5.2
Conjugate Alexa Fluor 647
Description Cytokeratin-7/-8; CK-7/CK-8;
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human HT29 colorectal carcinoma cell line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.