Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin Epithelial Mouse anti-Bovine, Chicken, Human, Mouse, Rat, Rabbit, Clone: AE1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1612
Description
This Anti-Cytokeratin Epithelial Antibody, clone AE1 is validated for use in immunohistochemistry (paraffin) for the detection of Cytokeratin Epithelial.
Specifications
| Cytokeratin Epithelial | |
| Monoclonal | |
| Unconjugated | |
| NM_002273.2 | |
| Mouse | |
| 500 μg | |
| Bovine, Chicken, Human, Mouse, Rabbit, Rat | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| AE1 | |
| Liquid in 0.05M Borate Buffered Saline, 0.15M NaCl, 0.09% Sodium Azide, pH8.2. | |
| KRT8, KO, K8, CYK8, Keratin-8, CK-8, Cytokeratin-8, K2C8, CK8, CARD2 | |
| Human epidermal keratin | |
| Primary | |
| 2-8°C in undiluted aliquots for up to 6 months. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction