Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin Mouse anti-Human, PE, Clone: CAM 5.2, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 347204
Description
The Anti-Cytokeratin antibody, clone CAM 5.2, is derived from the hybridization of P3/NS-1/1-Ag4-1 mouse myeloma cells with spleen cells from BALB/c mice immunized with the human colorectal carcinoma cell line HT29.

Specifications
Cytokeratin | |
Monoclonal | |
6μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Human HT29 colorectal carcinoma cell line | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
CAM 5.2 | |
PE | |
Mouse | |
50 Tests | |
Immunology | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction