Learn More
Cytomegalovirus gB Late Antigen, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA127221
Description
Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5). Glycoprotein B (gB) is the main antigenic protein within the envelope of HCMV. It is required for HCMV mediated membrane fusion along with the gH/gL complex. The unprocessed precursor of HCMV gB is 906 amino acids in length, with a MW of approximately 100kDa.
PA1-27221 detects Cytomegalovirus from virus samples. PA1-27221 will not react with HSV, VZV, HEp-2 cells or human fibroblasts .PA1-27221 has been successfully used in immunofluorescence, and Western blot procedures.The PA1-27221 immunogen is a cell preparation with purified cytomegalovirus virions of strain AD169.
Specifications
Cytomegalovirus gB Late Antigen | |
Polyclonal | |
Unconjugated | |
Goat | |
Purified | |
RUO | |
Virus | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
4mg/mL | |
CMV | |
Cell preparation with purified cytomegalovirus virions of strain AD169. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.