Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytomegalovirus Mouse anti-Human, Clone: 8B1.2, 1G5.2, and 2D4.2, MilliporeSigma™

Catalog No. MAB8121MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8121MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8121MI Supplier MilliporeSigma Supplier No. MAB8121
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Cytomegalovirus Antibody, blend, clone 8B1.2, 1G5.2 and 2D4.2 is validated for use in immunofluoresence, immunohistochemistry for the detection of Cytomegalovirus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytomegalovirus
Applications Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 8B1.2, 1G5.2, and 2D4.2, 1G5.2, and 2D4.2
Conjugate Unconjugated
Formulation 0.02MPB, 0.25M NaCl, PH 7.6 with 0.1% Sodium Azide.
Host Species Mouse
Immunogen MRC-5 infected cells with ATCC VR538 and AD169, ultrasonicated, screened by plate reduction neutralization tests.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8C in convenient aliquots for up to 6 months. Do not Freeze.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.