Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytomegalovirus Mouse anti-Human, Clone: 8B1.2, MilliporeSigma™

Catalog No. MAB810R500U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB810R500U 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB810R500U Supplier MilliporeSigma Supplier No. MAB810R500UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Cytomegalovirus Antibody, clone 8B1.2 is an antibody against Cytomegalovirus for use in ELISA, flow cytometry, immunofluoresence, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytomegalovirus
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8B1.2
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Host Species Mouse
Immunogen Affinity purified immediate early antigen from MRC-5 cells infected with CMV AD169 (ATCC).
Purification Method Protein A purfied
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.